Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCAGGCATATGCCCGTGTTCCT[C/T]TCTTCTTTATCAACTTTGCTTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614477 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf37 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8orf37 - chromosome 8 open reading frame 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177965.3 | 1838 | Missense Mutation | AAA,AGA | K,R 174 | NP_808880.1 | |
XM_005250799.3 | 1838 | Missense Mutation | AAA,AGA | K,R 255 | XP_005250856.2 |