Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATATTCATTGGCATGTGGTAT[A/G]TGGTAGTATTTTACTGCCGGGATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606140 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XPO7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XPO7 - exportin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015024.4 | Intron | NP_055839.3 | ||||
XM_017013244.1 | Intron | XP_016868733.1 | ||||
XM_017013245.1 | Intron | XP_016868734.1 | ||||
XM_017013246.1 | Intron | XP_016868735.1 | ||||
XM_017013247.1 | Intron | XP_016868736.1 |