Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTCAATGTCACGCACCCTAGC[A/T]GTGTGCATCTTCATGAGTTCTTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601060 | ||||||||||||||||||||
Literature Links: |
ENPP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENPP2 - ectonucleotide pyrophosphatase/phosphodiesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040092.2 | 2558 | Silent Mutation | ACA,ACT | T,T 824 | NP_001035181.1 | |
NM_001130863.2 | 2558 | Silent Mutation | ACA,ACT | T,T 849 | NP_001124335.1 | |
NM_006209.4 | 2558 | Silent Mutation | ACA,ACT | T,T 876 | NP_006200.3 | |
XM_006716584.1 | 2558 | Silent Mutation | ACA,ACT | T,T 901 | XP_006716647.1 | |
XM_006716585.1 | 2558 | Silent Mutation | ACA,ACT | T,T 897 | XP_006716648.1 | |
XM_006716587.1 | 2558 | Silent Mutation | ACA,ACT | T,T 872 | XP_006716650.1 | |
XM_017013571.1 | 2558 | Silent Mutation | ACA,ACT | T,T 845 | XP_016869060.1 | |
XM_017013572.1 | 2558 | Silent Mutation | ACA,ACT | T,T 845 | XP_016869061.1 | |
XM_017013573.1 | 2558 | Silent Mutation | ACA,ACT | T,T 841 | XP_016869062.1 | |
XM_017013574.1 | 2558 | Silent Mutation | ACA,ACT | T,T 820 | XP_016869063.1 | |
XM_017013575.1 | 2558 | Silent Mutation | ACA,ACT | T,T 820 | XP_016869064.1 |