Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAGTCGTGGACGTACATGAGGAC[A/G]GCTATTGGCTGTCCGATGATGAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604900 MIM: 140580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGAT1 - diacylglycerol O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012079.5 | 1408 | Silent Mutation | GCC,GCT | A,A 468 | NP_036211.2 | |
XM_011517356.2 | 1408 | Silent Mutation | GCC,GCT | A,A 411 | XP_011515658.1 |
HSF1 - heat shock transcription factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6848 - microRNA 6848 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |