Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTAGGGACCCTTCACATTACCC[A/G]GCCGGGCAGCAAGGCTGGCGAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610026 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL22A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL22A1 - collagen type XXII alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152888.2 | 3458 | Missense Mutation | CCG,CTG | P,L 1620 | NP_690848.1 | |
XM_011516883.2 | 3458 | Missense Mutation | CCG,CTG | P,L 1600 | XP_011515185.1 | |
XM_011516884.2 | 3458 | Missense Mutation | CCG,CTG | P,L 1587 | XP_011515186.1 | |
XM_011516885.2 | 3458 | Missense Mutation | CCG,CTG | P,L 1582 | XP_011515187.1 | |
XM_011516886.2 | 3458 | Missense Mutation | CCG,CTG | P,L 1571 | XP_011515188.1 | |
XM_011516887.2 | 3458 | Missense Mutation | CCG,CTG | P,L 1258 | XP_011515189.1 | |
XM_011516888.2 | 3458 | Intron | XP_011515190.1 | |||
XM_011516889.2 | 3458 | Missense Mutation | CCG,CTG | P,L 1052 | XP_011515191.1 | |
XM_017013150.1 | 3458 | Missense Mutation | CCG,CTG | P,L 1558 | XP_016868639.1 | |
XM_017013151.1 | 3458 | Missense Mutation | CCG,CTG | P,L 1546 | XP_016868640.1 | |
XM_017013152.1 | 3458 | Missense Mutation | CCG,CTG | P,L 1258 | XP_016868641.1 |