Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATGAGCCACCATGCCTGGGCTTT[A/C]ATGATAGGTTTTCAAACTCCACATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTFR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTFR1 - mitochondrial fission regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145838.1 | Intron | NP_001139310.1 | ||||
NM_001145839.1 | Intron | NP_001139311.1 | ||||
NM_014637.3 | Intron | NP_055452.3 | ||||
XM_006716484.2 | Intron | XP_006716547.2 | ||||
XM_011517626.1 | Intron | XP_011515928.1 | ||||
XM_011517627.2 | Intron | XP_011515929.1 | ||||
XM_011517628.2 | Intron | XP_011515930.1 |