Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACATAGATTTCTTAGAGTGGGGC[C/T]AAGGAAATACACATATATTTTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602419 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EGR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EGR3 - early growth response 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199880.1 | 2369 | UTR 3 | NP_001186809.1 | |||
NM_001199881.1 | 2369 | UTR 3 | NP_001186810.1 | |||
NM_004430.2 | 2369 | UTR 3 | NP_004421.2 | |||
XM_005273425.3 | 2369 | UTR 3 | XP_005273482.1 | |||
XM_005273426.3 | 2369 | UTR 3 | XP_005273483.1 | |||
XM_011544429.2 | 2369 | UTR 3 | XP_011542731.1 |