Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGCTTTCAGGAGCGCCTCCCCA[A/G]GGAGAAGGTGCCCCGGGGCTCCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611083 | ||||||||||||||||||||
Literature Links: |
ADHFE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADHFE1 - alcohol dehydrogenase, iron containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RRS1 - ribosome biogenesis regulator homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015169.3 | 826 | Missense Mutation | AAG,AGG | K,R 241 | NP_055984.1 |
RRS1-AS1 - RRS1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |