Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACTCTGGATTTCAGTGTGGCGA[C/T]TTCTCCCTTATAGGGTGCACGCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613326 | ||||||||||||||||||||
Literature Links: |
DPYS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPYS - dihydropyrimidinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001385.2 | 1592 | Missense Mutation | ATC,GTC | I,V 497 | NP_001376.1 | |
XM_005250818.3 | 1592 | Missense Mutation | ATC,GTC | I,V 533 | XP_005250875.1 | |
XM_006716518.3 | 1592 | Missense Mutation | ATC,GTC | I,V 480 | XP_006716581.1 | |
XM_011516903.2 | 1592 | Intron | XP_011515205.1 | |||
XM_017013167.1 | 1592 | Intron | XP_016868656.1 |