Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCCTCGGTGGAGGCCCAGTACA[C/T]CCAGGCCCAGGCCAGGGAGCCCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM83A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM83A - family with sequence similarity 83 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288587.2 | 1141 | Missense Mutation | ACC,ATC | T,I 71 | NP_001275516.1 | |
NM_001321630.1 | 1141 | Missense Mutation | ACC,ATC | T,I 71 | NP_001308559.1 | |
NM_032899.6 | 1141 | Missense Mutation | ACC,ATC | T,I 71 | NP_116288.2 | |
NM_207006.2 | 1141 | Missense Mutation | ACC,ATC | T,I 71 | NP_996889.1 | |
XM_005251087.2 | 1141 | Missense Mutation | ACC,ATC | T,I 71 | XP_005251144.1 | |
XM_011517338.1 | 1141 | Missense Mutation | ACC,ATC | T,I 71 | XP_011515640.1 |