Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCCGGACTTGGTGCAGAAAAAA[C/T]ACCAGCCGTGCTGTTAAACCAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191305 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BLK - BLK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001715.2 | 155 | UTR 5 | NP_001706.2 | |||
XM_011543824.1 | 155 | UTR 5 | XP_011542126.1 | |||
XM_011543825.2 | 155 | Intron | XP_011542127.1 | |||
XM_011543827.1 | 155 | UTR 5 | XP_011542129.1 | |||
XM_011543828.2 | 155 | UTR 5 | XP_011542130.1 | |||
XM_011543829.2 | 155 | UTR 5 | XP_011542131.1 | |||
XM_017013730.1 | 155 | UTR 5 | XP_016869219.1 |