Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACCAGCCCTGTGTCGTGCAAAA[A/T]GCATTCATCGCCCTGCCCCTGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMBOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMBOX1 - homeobox containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIF13B - kinesin family member 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015254.3 | 5708 | UTR 3 | NP_056069.2 | |||
XM_005273458.1 | 5708 | UTR 3 | XP_005273515.1 | |||
XM_011544457.1 | 5708 | UTR 3 | XP_011542759.1 | |||
XM_011544458.1 | 5708 | UTR 3 | XP_011542760.1 | |||
XM_011544459.2 | 5708 | UTR 3 | XP_011542761.1 | |||
XM_011544460.2 | 5708 | UTR 3 | XP_011542762.1 | |||
XM_011544461.1 | 5708 | UTR 3 | XP_011542763.1 | |||
XM_017013257.1 | 5708 | Intron | XP_016868746.1 |