Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTGAGAAAAAGTGTCGCTTAAC[A/G]AAGTCACAGGAGGAGGCACACATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603625 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LY6H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LY6H - lymphocyte antigen 6 complex, locus H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130478.1 | 403 | Silent Mutation | TTC,TTT | F,F 100 | NP_001123950.1 | |
NM_001135655.1 | 403 | Silent Mutation | TTC,TTT | F,F 100 | NP_001129127.1 | |
NM_002347.4 | 403 | Silent Mutation | TTC,TTT | F,F 79 | NP_002338.3 | |
XM_017013413.1 | 403 | Silent Mutation | TTC,TTT | F,F 105 | XP_016868902.1 | |
XM_017013414.1 | 403 | Silent Mutation | TTC,TTT | F,F 79 | XP_016868903.1 |