Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTATTTAAATTTTGTACCTGCCT[A/T]GCAACAGTATTCTCATTCCTAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605772 | ||||||||||||||||||||
Literature Links: |
EBAG9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EBAG9 - estrogen receptor binding site associated, antigen, 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278938.1 | 280 | Silent Mutation | CTA,CTT | L,L 15 | NP_001265867.1 | |
NM_004215.4 | 280 | Silent Mutation | CTA,CTT | L,L 15 | NP_004206.1 | |
NM_198120.2 | 280 | Silent Mutation | CTA,CTT | L,L 15 | NP_936056.1 | |
XM_017013960.1 | 280 | Silent Mutation | CTA,CTT | L,L 15 | XP_016869449.1 |