Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCTGCTGCTTGGTCTTCTGCTG[C/G]TGCACGAAGCGCTGGAGGATGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604584 MIM: 601872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDGFRL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDGFRL - platelet derived growth factor receptor like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006207.2 | 479 | Missense Mutation | CTG,GTG | L,V 12 | NP_006198.1 |
SLC7A2 - solute carrier family 7 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |