Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATTCCAGAAGTTCTAATTACTG[C/T]AATTAACTGGGATGCTTTCTAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604386 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPS1 - transcriptional repressor GATA binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282902.2 | 8770 | UTR 3 | NP_001269831.1 | |||
NM_001282903.2 | 8770 | UTR 3 | NP_001269832.1 | |||
NM_014112.4 | 8770 | UTR 3 | NP_054831.2 | |||
XM_005251049.2 | 8770 | UTR 3 | XP_005251106.1 | |||
XM_006716625.1 | 8770 | UTR 3 | XP_006716688.1 | |||
XM_011517264.1 | 8770 | UTR 3 | XP_011515566.1 | |||
XM_011517266.2 | 8770 | UTR 3 | XP_011515568.1 | |||
XM_011517267.1 | 8770 | UTR 3 | XP_011515569.1 | |||
XM_011517268.1 | 8770 | UTR 3 | XP_011515570.1 | |||
XM_017013801.1 | 8770 | UTR 3 | XP_016869290.1 |