Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGCAAACTTTGAGAAGAACGTGA[A/G]CCAAGCTATCCACAAGTACAATGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603258 MIM: 174760 | ||||||||||||||||||||
Literature Links: |
IKBKB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IKBKB - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLB - DNA polymerase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002690.2 | 256 | Missense Mutation | AAC,AGC | N,S 30 | NP_002681.1 | |
XM_005273535.3 | 256 | Missense Mutation | AAC,AGC | N,S 30 | XP_005273592.1 | |
XM_005273536.3 | 256 | Missense Mutation | AAC,AGC | N,S 30 | XP_005273593.1 | |
XM_005273537.3 | 256 | Missense Mutation | AAC,AGC | N,S 30 | XP_005273594.1 | |
XM_005273538.2 | 256 | Intron | XP_005273595.1 | |||
XM_005273539.2 | 256 | Intron | XP_005273596.1 | |||
XM_005273540.4 | 256 | Intron | XP_005273597.1 | |||
XM_006716353.2 | 256 | UTR 5 | XP_006716416.1 | |||
XM_017013583.1 | 256 | Intron | XP_016869072.1 | |||
XM_017013584.1 | 256 | Intron | XP_016869073.1 |