Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTCCTTGCTGGTGCTGTCCTC[A/G]TGGAACCGGATGTGGAGGAAGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611966 | ||||||||||||||||||||
Literature Links: |
TRAPPC9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRAPPC9 - trafficking protein particle complex 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160372.3 | 3645 | Silent Mutation | CAC,CAT | H,H 1121 | NP_001153844.1 | |
NM_001321646.1 | 3645 | Silent Mutation | CAC,CAT | H,H 1112 | NP_001308575.1 | |
NM_031466.7 | 3645 | Silent Mutation | CAC,CAT | H,H 1219 | NP_113654.4 | |
XM_011517326.2 | 3645 | Silent Mutation | CAC,CAT | H,H 1210 | XP_011515628.1 | |
XM_011517328.2 | 3645 | Intron | XP_011515630.1 | |||
XM_011517330.2 | 3645 | Silent Mutation | CAC,CAT | H,H 604 | XP_011515632.1 | |
XM_017013893.1 | 3645 | Intron | XP_016869382.1 | |||
XM_017013894.1 | 3645 | Silent Mutation | CAC,CAT | H,H 661 | XP_016869383.1 |