Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCTCGAGTGTCTATCCACCAGC[A/G]CTAAACCCTACAAGAAATGAGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613203 MIM: 604912 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSCC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DSCC1 - DNA replication and sister chromatid cohesion 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024094.2 | 1067 | Missense Mutation | GCG,GTG | A,V 311 | NP_076999.2 | |
XM_005251065.3 | 1067 | Missense Mutation | GCG,GTG | A,V 266 | XP_005251122.1 |
TAF2 - TATA-box binding protein associated factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |