Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCATCCCAACATCATCAGCCTC[C/T]TTGACGTGATCCGGGCAGAGAACGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611927 | ||||||||||||||||||||
Literature Links: |
FAM83H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM83H - family with sequence similarity 83 member H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928160 - uncharacterized LOC101928160 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK15 - mitogen-activated protein kinase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139021.2 | 1108 | Missense Mutation | CTT,TTT | L,F 77 | NP_620590.2 | |
XM_011516925.2 | 1108 | Missense Mutation | CTT,TTT | L,F 98 | XP_011515227.1 | |
XM_011516926.2 | 1108 | Missense Mutation | CTT,TTT | L,F 96 | XP_011515228.1 | |
XM_011516927.2 | 1108 | Missense Mutation | CTT,TTT | L,F 79 | XP_011515229.1 | |
XM_011516928.2 | 1108 | Missense Mutation | CTT,TTT | L,F 98 | XP_011515230.1 | |
XM_017013218.1 | 1108 | Missense Mutation | CTT,TTT | L,F 98 | XP_016868707.1 |