Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTTCCTAAATAAAGGAACATAA[A/C]TGTACACTAAAGGGTGTTTCCCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611990 MIM: 604068 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPS28 - mitochondrial ribosomal protein S28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPD52 - tumor protein D52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025252.2 | 972 | UTR 3 | NP_001020423.1 | |||
NM_001025253.2 | 972 | UTR 3 | NP_001020424.1 | |||
NM_001287140.1 | 972 | UTR 3 | NP_001274069.1 | |||
NM_001287142.1 | 972 | UTR 3 | NP_001274071.1 | |||
NM_001287143.1 | 972 | UTR 3 | NP_001274072.1 | |||
NM_001287144.1 | 972 | UTR 3 | NP_001274073.1 | |||
NM_005079.3 | 972 | UTR 3 | NP_005070.1 |