Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGATGATACTTTCTCGTATGTTC[A/G]TGTACAGATAACACTGTTCACAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FABP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FABP12 - fatty acid binding protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105281.1 | 379 | Nonsense Mutation | CGA,TGA | R,* 127 | NP_001098751.1 | |
XM_005251285.4 | 379 | Nonsense Mutation | CGA,TGA | R,* 127 | XP_005251342.1 | |
XM_006716465.3 | 379 | Nonsense Mutation | CGA,TGA | R,* 127 | XP_006716528.1 | |
XM_011517577.2 | 379 | Nonsense Mutation | CGA,TGA | R,* 127 | XP_011515879.1 |