Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGCCTCCGCAGCCCCAGCCGAG[A/G]CAGCCCCCGCCGCCAGCGCCGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBTB10 - zinc finger and BTB domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105539.2 | 733 | Silent Mutation | AGA,AGG | R,R 45 | NP_001099009.1 | |
NM_001277145.1 | 733 | Intron | NP_001264074.1 | |||
NM_023929.4 | 733 | Silent Mutation | AGA,AGG | R,R 45 | NP_076418.3 |