Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGCAGCACTGGCAGTGCTGGCA[A/G]TGACAGCAGCACCAGCGGGAGTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612974 | ||||||||||||||||||||
Literature Links: |
DEPTOR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DEPTOR - DEP domain containing MTOR-interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283012.1 | 227 | Missense Mutation | AAT,AGT | N,S 12 | NP_001269941.1 | |
NM_022783.3 | 227 | Missense Mutation | AAT,AGT | N,S 12 | NP_073620.2 | |
XM_017013735.1 | 227 | Intron | XP_016869224.1 |