Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTGTGTTCCCAAGTAGTGGCT[A/G]TAGAAATAGCCCTGCAGTTCCAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606888 | ||||||||||||||||||||
Literature Links: |
CHRNA6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNA6 - cholinergic receptor nicotinic alpha 6 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199279.1 | 1809 | Nonsense Mutation | CAG,TAG | Q,* 470 | NP_001186208.1 | |
NM_004198.3 | 1809 | Nonsense Mutation | CAG,TAG | Q,* 485 | NP_004189.1 |