Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCGTGGCCAAAGCCAAAACCAC[A/G]TGATTTCCCTTTCAGCATTTATACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614426 | ||||||||||||||||||||
Literature Links: |
MAK16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAK16 - MAK16 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032509.3 | 2646 | Silent Mutation | ACA,ACG | T,T 300 | NP_115898.2 |
TTI2 - TELO2 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102401.2 | 2646 | UTR 3 | NP_001095871.1 | |||
NM_001265581.1 | 2646 | UTR 3 | NP_001252510.1 | |||
NM_025115.3 | 2646 | UTR 3 | NP_079391.2 | |||
XM_011544665.2 | 2646 | Intron | XP_011542967.1 |