Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTTCACCTGTGAACCAATTACT[G/T]TTCCCAGATGTATGAAAATGGCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603409 | ||||||||||||||||||||
Literature Links: |
FZD6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FZD6 - frizzled class receptor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164615.1 | 402 | Missense Mutation | GTT,TTT | V,F 29 | NP_001158087.1 | |
NM_001164616.1 | 402 | Intron | NP_001158088.1 | |||
NM_001317796.1 | 402 | UTR 5 | NP_001304725.1 | |||
NM_003506.3 | 402 | Missense Mutation | GTT,TTT | V,F 29 | NP_003497.2 |