Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTTGGAGTGAGGAACATCAGCA[C/G]CCCTCGGGGCAGGCACAGCATCACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603937 | ||||||||||||||||||||
Literature Links: |
RP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RP1 - retinitis pigmentosa 1 (autosomal dominant) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006269.1 | 1565 | Missense Mutation | ACC,AGC | T,S 85 | NP_006260.1 | |
XM_017013721.1 | 1565 | Missense Mutation | ACC,AGC | T,S 92 | XP_016869210.1 | |
XM_017013722.1 | 1565 | Missense Mutation | ACC,AGC | T,S 85 | XP_016869211.1 |