Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCTGCAACCACCCAGTGATCAT[C/T]GGAGGGAAAAAAGCTTGACTTACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602747 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP4 - dual specificity phosphatase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001394.6 | 5362 | UTR 3 | NP_001385.1 | |||
NM_057158.3 | 5362 | UTR 3 | NP_476499.1 | |||
XM_011544428.2 | 5362 | UTR 3 | XP_011542730.1 |