Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCTCAGTGACACCGGAAGGAAC[A/G]TCCGAGTCAGAAGTGTGGGAAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185430 MIM: 132811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLU PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLU - clusterin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001831.3 | 3283 | Silent Mutation | GAC,GAT | D,D 397 | NP_001822.3 | |
XM_006716284.2 | 3283 | Silent Mutation | GAC,GAT | D,D 449 | XP_006716347.1 |
EPHX2 - epoxide hydrolase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |