Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCTGAAACATGAAACCAAACAG[A/G]CTTTGATATTTTTTTTTTTTTAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614426 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAK16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAK16 - MAK16 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032509.3 | 1613 | UTR 3 | NP_115898.2 |
TTI2 - TELO2 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102401.2 | 1613 | Intron | NP_001095871.1 | |||
NM_001265581.1 | 1613 | Intron | NP_001252510.1 | |||
NM_025115.3 | 1613 | Intron | NP_079391.2 | |||
XM_011544665.2 | 1613 | Intron | XP_011542967.1 |