Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACCTTCATGTGTGACATCCAGC[C/T]GTCAGCAGGAAGAAGTCAGTCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609740 MIM: 601711 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHF19 - PHD finger protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009936.2 | 3557 | Intron | NP_001009936.1 | |||
NM_001286840.1 | 3557 | Intron | NP_001273769.1 | |||
NM_001286842.1 | 3557 | Intron | NP_001273771.1 | |||
NM_001286843.1 | 3557 | Intron | NP_001273772.1 | |||
NM_015651.2 | 3557 | Intron | NP_056466.1 |
TRAF1 - TNF receptor associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190945.1 | 3557 | UTR 3 | NP_001177874.1 | |||
NM_001190947.1 | 3557 | UTR 3 | NP_001177876.1 | |||
NM_005658.4 | 3557 | UTR 3 | NP_005649.1 |