Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCAGCCACATAAAAATCCACAC[G/T]ATTCCTTCCTGTGTTGGCCTGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605573 MIM: 609182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD17B3 - hydroxysteroid 17-beta dehydrogenase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35D2 - solute carrier family 35 member D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286990.1 | Intron | NP_001273919.1 | ||||
NM_007001.2 | Intron | NP_008932.2 | ||||
XM_005251673.1 | Intron | XP_005251730.1 | ||||
XM_005251674.1 | Intron | XP_005251731.1 | ||||
XM_005251675.2 | Intron | XP_005251732.1 | ||||
XM_005251676.1 | Intron | XP_005251733.1 | ||||
XM_005251678.4 | Intron | XP_005251735.1 | ||||
XM_006716939.2 | Intron | XP_006717002.1 | ||||
XM_011518164.1 | Intron | XP_011516466.1 |