Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCTTGGCCTCGCCTCTGGGCGTC[C/T]TCTGAGATTGCTGTCACCACCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600981 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01506 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01506 - long intergenic non-protein coding RNA 1506 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105376072 - uncharacterized LOC105376072 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGM5 - phosphoglucomutase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM252 - transmembrane protein 252 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153237.1 | 536 | Silent Mutation | GAA,GAG | E,E 161 | NP_694969.1 |