Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCCTTGACTGGAGACTGGATCC[A/C]CTTCAAACAGCTAGGTTTCCCTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611258 | ||||||||||||||||||||
Literature Links: |
TDRD7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TDRD7 - tudor domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302884.1 | 221 | Intron | NP_001289813.1 | |||
NM_014290.2 | 221 | Missense Mutation | CAC,CCC | H,P 40 | NP_055105.2 | |
XM_006717026.3 | 221 | UTR 5 | XP_006717089.1 |