Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGGGCTCTGATGAATGTAAAG[C/G]AGTGGGTTCAGAAGAAATAAACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605976 | ||||||||||||||||||||
Literature Links: |
ZBTB26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB26 - zinc finger and BTB domain containing 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304363.1 | 711 | Missense Mutation | CCT,GCT | P,A 210 | NP_001291292.1 | |
NM_001304364.1 | 711 | Missense Mutation | CCT,GCT | P,A 210 | NP_001291293.1 | |
NM_020924.3 | 711 | Missense Mutation | CCT,GCT | P,A 210 | NP_065975.1 |
ZBTB6 - zinc finger and BTB domain containing 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |