Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGTCCATTTACGCTGAGAATAG[C/T]TTGAAATCAGGTAAAAATAACCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116901 MIM: 607693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CKS2 - CDC28 protein kinase regulatory subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SECISBP2 - SECIS binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282688.1 | 722 | Silent Mutation | AGC,AGT | S,S 187 | NP_001269617.1 | |
NM_001282689.1 | 722 | Silent Mutation | AGC,AGT | S,S 115 | NP_001269618.1 | |
NM_001282690.1 | 722 | Silent Mutation | AGC,AGT | S,S 120 | NP_001269619.1 | |
NM_024077.4 | 722 | Silent Mutation | AGC,AGT | S,S 188 | NP_076982.3 | |
XM_005252196.2 | 722 | Silent Mutation | AGC,AGT | S,S 188 | XP_005252253.1 | |
XM_005252202.3 | 722 | Intron | XP_005252259.1 | |||
XM_006717282.2 | 722 | Silent Mutation | AGC,AGT | S,S 115 | XP_006717345.1 | |
XM_011519000.2 | 722 | Silent Mutation | AGC,AGT | S,S 120 | XP_011517302.1 | |
XM_011519001.1 | 722 | Silent Mutation | AGC,AGT | S,S 120 | XP_011517303.1 | |
XM_011519002.1 | 722 | Silent Mutation | AGC,AGT | S,S 120 | XP_011517304.1 | |
XM_011519003.1 | 722 | Silent Mutation | AGC,AGT | S,S 120 | XP_011517305.1 | |
XM_017015122.1 | 722 | Silent Mutation | AGC,AGT | S,S 188 | XP_016870611.1 | |
XM_017015123.1 | 722 | Silent Mutation | AGC,AGT | S,S 188 | XP_016870612.1 | |
XM_017015124.1 | 722 | Silent Mutation | AGC,AGT | S,S 188 | XP_016870613.1 | |
XM_017015125.1 | 722 | UTR 5 | XP_016870614.1 |