Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCGAGGATCATGGTGAACCAGCC[A/G]GTGCTGAGGAAGGAGCCCGACTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606378 MIM: 610135 | ||||||||||||||||||||
Literature Links: |
ST6GALNAC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ST6GALNAC4 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175039.3 | 1038 | Silent Mutation | NP_778204.1 | |||
NM_175040.3 | 1038 | Silent Mutation | NP_778205.1 | |||
XM_011518527.2 | 1038 | Silent Mutation | XP_011516829.1 | |||
XM_011518529.2 | 1038 | Missense Mutation | XP_011516831.1 |
ST6GALNAC6 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |