Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTAGGTACTGTATTGTTATTATG[C/T]ATGGTTAAAGTGGCAAAGGTTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RLN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RLN1 - relaxin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006911.3 | 974 | UTR 3 | NP_008842.1 | |||
XM_017015007.1 | 974 | Intron | XP_016870496.1 | |||
XM_017015008.1 | 974 | Intron | XP_016870497.1 | |||
XM_017015009.1 | 974 | Intron | XP_016870498.1 | |||
XM_017015010.1 | 974 | Intron | XP_016870499.1 |