Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTATCACAATTTGTAACTGCTT[C/T]ATTTCTTTGTTTTCTTCTTAGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125270 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALAD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALAD - aminolevulinate dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000031.5 | 2174 | UTR 3 | NP_000022.3 | |||
NM_001003945.2 | 2174 | UTR 3 | NP_001003945.1 | |||
NM_001317745.1 | 2174 | UTR 3 | NP_001304674.1 | |||
XM_011518364.2 | 2174 | UTR 3 | XP_011516666.1 |