Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGCAGGCGCTCCAGCAGCTGCC[A/G]GTAGTTCCTCCCGGTCAGCGGCGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 185641 MIM: 185642 | ||||||||||||||||||||
Literature Links: |
MED22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED22 - mediator complex subunit 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SURF6 - surfeit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278942.1 | 787 | UTR 3 | NP_001265871.1 | |||
NM_006753.5 | 787 | Missense Mutation | CGG,TGG | R,W 237 | NP_006744.2 |