Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGACCACCGGCTGCGCCTGTGG[A/G]GACTCTTTCAGACGCTCGACGTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610673 MIM: 608745 | ||||||||||||||||||||
Literature Links: |
NAIF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAIF1 - nuclear apoptosis inducing factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_197956.3 | 274 | Intron | NP_931045.1 |
SLC25A25 - solute carrier family 25 member 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006641.3 | 274 | Missense Mutation | AGA,GGA | R,G 52 | NP_001006642.1 | |
NM_001006642.3 | 274 | Intron | NP_001006643.1 | |||
NM_001265614.2 | 274 | Intron | NP_001252543.1 | |||
NM_052901.4 | 274 | Intron | NP_443133.2 | |||
XM_005251688.1 | 274 | Missense Mutation | AGA,GGA | R,G 52 | XP_005251745.1 | |
XM_005251689.4 | 274 | Intron | XP_005251746.1 | |||
XM_006716948.2 | 274 | Intron | XP_006717011.1 | |||
XM_006716949.3 | 274 | Intron | XP_006717012.1 |