Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGCCCTGTGGAGGGTGGGGGTG[A/C]AGGGTTGGTTGGACCCTGGGAAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607387 | ||||||||||||||||||||
Literature Links: |
ARID3C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARID3C - AT-rich interaction domain 3C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017363.1 | 4281 | Missense Mutation | GCA,TCA | A,S 399 | NP_001017363.1 | |
XM_017014283.1 | 4281 | Missense Mutation | GCA,TCA | A,S 549 | XP_016869772.1 |
DCTN3 - dynactin subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281425.1 | 4281 | Intron | NP_001268354.1 | |||
NM_001281426.1 | 4281 | Intron | NP_001268355.1 | |||
NM_001281427.1 | 4281 | Intron | NP_001268356.1 | |||
NM_007234.4 | 4281 | Intron | NP_009165.1 | |||
NM_024348.3 | 4281 | Intron | NP_077324.1 |
RPP25L - ribonuclease P/MRP subunit p25 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |