Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAGATAGGGCACTGTTTCCACG[C/T]GGCTCTGGATGTAGGCACTCCGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612856 | ||||||||||||||||||||
Literature Links: |
ASTN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASTN2 - astrotactin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184734.1 | 1258 | Missense Mutation | NP_001171663.1 | |||
NM_001184735.1 | 1258 | Intron | NP_001171664.1 | |||
NM_014010.4 | 1258 | Missense Mutation | NP_054729.3 | |||
NM_198186.3 | 1258 | Missense Mutation | NP_937829.3 | |||
NM_198187.3 | 1258 | Missense Mutation | NP_937830.3 | |||
NM_198188.2 | 1258 | Missense Mutation | NP_937831.1 |