Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGATGACATCAGCTCCCTGGAGA[A/G]TCAGCCCCTGAAAGTTGGAATTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605284 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPACA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPACA9 - sperm acrosome associated 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSC1 - tuberous sclerosis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000368.4 | 4901 | UTR 3 | NP_000359.1 | |||
NM_001162426.1 | 4901 | UTR 3 | NP_001155898.1 | |||
NM_001162427.1 | 4901 | UTR 3 | NP_001155899.1 | |||
XM_005272211.1 | 4901 | UTR 3 | XP_005272268.1 | |||
XM_006717271.1 | 4901 | UTR 3 | XP_006717334.1 | |||
XM_011518979.2 | 4901 | UTR 3 | XP_011517281.1 | |||
XM_017015096.1 | 4901 | UTR 3 | XP_016870585.1 | |||
XM_017015097.1 | 4901 | UTR 3 | XP_016870586.1 | |||
XM_017015098.1 | 4901 | UTR 3 | XP_016870587.1 | |||
XM_017015099.1 | 4901 | UTR 3 | XP_016870588.1 | |||
XM_017015100.1 | 4901 | UTR 3 | XP_016870589.1 | |||
XM_017015101.1 | 4901 | UTR 3 | XP_016870590.1 |