Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCATGGTGACTGGTACACATCCC[C/T]GAGCAGCACTTGGGTGGAAAAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602012 MIM: 605798 | ||||||||||||||||||||
Literature Links: |
ENTPD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENTPD2 - ectonucleoside triphosphate diphosphohydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001246.3 | 883 | Missense Mutation | AGG,GGG | R,G 277 | NP_001237.1 | |
NM_203468.2 | 883 | Missense Mutation | AGG,GGG | R,G 277 | NP_982293.1 | |
XM_011519212.2 | 883 | Missense Mutation | AGG,GGG | R,G 174 | XP_011517514.1 |
NPDC1 - neural proliferation, differentiation and control 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |