Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGATTGCATTACATACCTGAGC[A/G]TAACAAGCTTCTTCTATGGCTAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600529 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AUH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AUH - AU RNA binding protein/enoyl-CoA hydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306190.1 | 1040 | Silent Mutation | TAC,TAT | Y,Y 283 | NP_001293119.1 | |
NM_001698.2 | 1040 | Silent Mutation | TAC,TAT | Y,Y 312 | NP_001689.1 | |
XM_005252066.3 | 1040 | Silent Mutation | TAC,TAT | Y,Y 322 | XP_005252123.1 | |
XM_005252067.4 | 1040 | Intron | XP_005252124.1 | |||
XM_005252069.4 | 1040 | Intron | XP_005252126.1 | |||
XM_005252072.2 | 1040 | Intron | XP_005252129.1 | |||
XM_006717150.3 | 1040 | Silent Mutation | TAC,TAT | Y,Y 293 | XP_006717213.1 | |
XM_011518800.2 | 1040 | Intron | XP_011517102.1 | |||
XM_011518802.2 | 1040 | Silent Mutation | TAC,TAT | Y,Y 203 | XP_011517104.1 | |
XM_011518803.2 | 1040 | Intron | XP_011517105.1 | |||
XM_011518804.2 | 1040 | Intron | XP_011517106.1 | |||
XM_017014849.1 | 1040 | Intron | XP_016870338.1 | |||
XM_017014850.1 | 1040 | Silent Mutation | TAC,TAT | Y,Y 203 | XP_016870339.1 | |
XM_017014851.1 | 1040 | Intron | XP_016870340.1 |