Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTTTAATCAGAATGAAGTGGGC[A/G]TTTTCTAGACTCACCATGAAAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604303 MIM: 603722 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTL7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTL7A - actin like 7A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IKBKAP - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318360.1 | 5280 | UTR 3 | NP_001305289.1 | |||
NM_003640.4 | 5280 | UTR 3 | NP_003631.2 | |||
XM_011519136.1 | 5280 | UTR 3 | XP_011517438.1 | |||
XM_011519137.1 | 5280 | UTR 3 | XP_011517439.1 | |||
XM_017015238.1 | 5280 | Intron | XP_016870727.1 |