Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATACATAGGGCCAAATCTTGTTG[A/G]TTTCTGTTCCGGAGAATGTTTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609290 MIM: 610346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK3 - adenylate kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199852.1 | 596 | Missense Mutation | ACC,ATC | T,I 161 | NP_001186781.1 | |
NM_001199853.1 | 596 | Missense Mutation | ACC,ATC | T,I 131 | NP_001186782.1 | |
NM_001199855.1 | 596 | Missense Mutation | ACC,ATC | T,I 131 | NP_001186784.1 | |
NM_001199856.1 | 596 | Missense Mutation | ACC,ATC | T,I 131 | NP_001186785.1 | |
NM_016282.3 | 596 | Missense Mutation | ACC,ATC | T,I 201 | NP_057366.2 |
CDC37L1 - cell division cycle 37 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |