Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATGCAGCTGAACTTCAGGAATT[C/T]TAGCATCCATTTAAAAAGATATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120900 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5 - complement component 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317163.1 | 5215 | Silent Mutation | TAA,TAG | *,* 1683 | NP_001304092.1 | |
NM_001317164.1 | 5215 | Intron | NP_001304093.1 | |||
NM_001735.2 | 5215 | Silent Mutation | TAA,TAG | *,* 1677 | NP_001726.2 | |
XM_011518980.2 | 5215 | Silent Mutation | TAA,TAG | *,* 1682 | XP_011517282.1 | |
XM_017015102.1 | 5215 | Silent Mutation | TAA,TAG | *,* 1682 | XP_016870591.1 | |
XM_017015103.1 | 5215 | Silent Mutation | TAA,TAG | *,* 1561 | XP_016870592.1 | |
XM_017015104.1 | 5215 | Intron | XP_016870593.1 |